Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D4
Entrez Gene ID
9882
Gene Name
TBC1 domain family member 4
Gene Aliases
AS160, NIDDM5
Chromosome Location
Chr.13: 75284664 - 75482646 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286658.2 NP_001273587.1 13 - 14 2971 144
NM_001286659.2 NP_001273588.1 12 - 13 2806 144
NM_014832.4 NP_055647.2 14 - 15 2995 144
XM_005266603.1 XP_005266660.1 13 - 14 3398 144
XM_005266605.2 XP_005266662.1 14 - 15 2323 144
XM_006719903.3 XP_006719966.1 14 - 15 2683 144
XM_011535331.1 XP_011533633.1 13 - 14 3362 144
XM_017020882.1 XP_016876371.1 12 - 13 2015 144
XM_017020883.1 XP_016876372.1 11 - 12 2009 144
XM_017020884.1 XP_016876373.1 2 - 3 1581 144
GenBank mRNA AK300468.1 - 3 - 4 504 144
AY211910.1 - 3 - 4 566 144
FM207106.1 - 12 - 13 2405 144
FM207107.1 - 13 - 14 2570 144

View Full Product Details