The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286658.2 | NP_001273587.1 | 19 - 20 | 4042 | 134 |
NM_001286659.2 | NP_001273588.1 | 18 - 19 | 3877 | 134 | |
NM_014832.4 | NP_055647.2 | 20 - 21 | 4066 | 134 | |
XM_005266603.1 | XP_005266660.1 | 19 - 20 | 4469 | 134 | |
XM_005266605.2 | XP_005266662.1 | 20 - 21 | 3394 | 134 | |
XM_006719903.3 | XP_006719966.1 | 20 - 21 | 3754 | 134 | |
XM_011535331.1 | XP_011533633.1 | 19 - 20 | 4433 | 134 | |
XM_017020882.1 | XP_016876371.1 | 18 - 19 | 3086 | 134 | |
XM_017020883.1 | XP_016876372.1 | 17 - 18 | 3080 | 134 | |
XM_017020884.1 | XP_016876373.1 | 8 - 9 | 2652 | 134 | |
GenBank mRNA | AB011175.1 | - | 21 - 22 | 4015 | 134 |
AB449885.1 | - | 21 - 22 | 3668 | 134 | |
AK300468.1 | - | 9 - 10 | 1575 | 134 | |
AK304091.1 | - | 8 - 9 | 1396 | 134 | |
BC151239.1 | - | 21 - 22 | 4007 | 134 | |
CR749622.1 | - | 10 - 11 | 1846 | 134 | |
FM207106.1 | - | 18 - 19 | 3476 | 134 | |
FM207107.1 | - | 19 - 20 | 3641 | 134 |