The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032454.1 | NP_115830.1 | 4 - 5 | 805 | 60 |
GenBank mRNA | BC143835.1 | - | 3 - 4 | 860 | 60 |
X77386.1 | - | 3 - 4 | 399 | 60 |