The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017677.3 | NP_060147.2 | 12 - 13 | 1575 | 75 |
GenBank mRNA | AK000133.1 | - | 8 - 9 | 895 | 75 |
AK225231.1 | - | 8 - 9 | 895 | 75 | |
BC012399.1 | - | 12 - 13 | 1553 | 75 |