The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002505.4 | NP_002496.1 | 7 - 8 | 894 | 87 |
NM_021705.3 | NP_068351.1 | 6 - 7 | 807 | 87 | |
GenBank mRNA | BC039244.1 | - | 6 - 7 | 1060 | 87 |
BM561405.1 | - | 6 - 7 | 88 | ||
GQ167198.1 | - | 6 - 7 | 713 | 87 | |
KU159426.1 | - | 6 - 7 | 851 | 87 | |
M59079.1 | - | 4 - 5 | 479 | 87 | |
X59711.1 | - | 7 - 8 | 890 | 87 |