The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289910.1 | NP_001276839.1 | 6 - 7 | 746 | 85 |
NM_001290114.1 | NP_001277043.1 | 4 - 5 | 721 | 85 | |
NM_002168.3 | NP_002159.2 | 6 - 7 | 979 | 85 | |
GenBank mRNA | AK222916.1 | - | 6 - 7 | 85 | |
AK294148.1 | - | 6 - 7 | 746 | 85 | |
AK299987.1 | - | 4 - 5 | 635 | 85 | |
AK312627.1 | - | 6 - 7 | 943 | 85 | |
AK316388.1 | - | 6 - 7 | 820 | 85 | |
BC009244.2 | - | 6 - 7 | 881 | 85 | |
BC071828.1 | - | 6 - 7 | 880 | 85 | |
X69433.1 | - | 6 - 7 | 901 | 85 |