The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173470.1 | NP_775741.1 | 3 - 4 | 538 | 100 |
GenBank mRNA | AK295829.1 | - | 4 - 5 | 515 | 100 |
AK312409.1 | - | 3 - 4 | 551 | 100 | |
AL157477.1 | - | 3 - 4 | 537 | 100 | |
BC033588.1 | - | 3 - 4 | 538 | 100 |