The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195124.1 | NP_001182053.1 | 6 - 7 | 878 | 125 |
NM_001195125.1 | NP_001182054.1 | 4 - 5 | 694 | 125 | |
NM_001256917.1 | NP_001243846.1 | 4 - 5 | 698 | 125 | |
GenBank mRNA | AK026130.1 | - | 4 - 5 | 690 | 125 |
BC015536.2 | - | 4 - 5 | 634 | 125 | |
BU839368.1 | - | 1 - 2 | 125 |