Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP11A
Entrez Gene ID
23250
Gene Name
ATPase phospholipid transporting 11A
Gene Aliases
ATPIH, ATPIS
Chromosome Location
Chr.13: 112690034 - 112887168 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015205.2 NP_056020.2 17 - 18 1895 110
NM_032189.3 NP_115565.3 17 - 18 1895 110
XM_005268299.4 XP_005268356.1 17 - 18 2190 110
XM_005268300.4 XP_005268357.1 17 - 18 2189 110
XM_005268301.4 XP_005268358.1 17 - 18 2190 110
XM_005268303.4 XP_005268360.1 17 - 18 2190 110
XM_005268305.4 XP_005268362.1 17 - 18 2189 110
XM_005268306.4 XP_005268363.1 17 - 18 2189 110
XM_011537480.2 XP_011535782.1 17 - 18 2190 110
XM_017020490.1 XP_016875979.1 17 - 18 2190 110
XM_017020491.1 XP_016875980.1 17 - 18 2190 110
XM_017020492.1 XP_016875981.1 17 - 18 2189 110
GenBank mRNA AB028944.2 - 16 - 17 1712 110
AK128241.1 - 16 - 17 1758 110
AL161996.1 - 1 - 2 69 110
BC024154.2 - 16 - 17 1731 110

View Full Product Details