Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLXNC1
Entrez Gene ID
10154
Gene Name
plexin C1
Gene Aliases
CD232, PLXN-C1, VESPR
Chromosome Location
Chr.12: 94148696 - 94307675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005761.2 NP_005752.1 15 - 16 3134 72
XM_006719186.3 XP_006719249.1 15 - 16 3161 72
XM_011537730.2 XP_011536032.1 15 - 16 3161 72
XM_011537731.2 XP_011536033.1 15 - 16 3161 72
XM_017018671.1 XP_016874160.1 15 - 16 3161 72
XM_017018672.1 XP_016874161.1 15 - 16 3161 72
GenBank mRNA AB208934.1 - 14 - 15 1815 72
AF030339.1 - 15 - 16 3134 72

View Full Product Details