The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005761.2 | NP_005752.1 | 27 - 28 | 4481 | 122 |
NR_037687.1 | 8 - 9 | 1025 | 122 | ||
XM_006719186.3 | XP_006719249.1 | 26 - 27 | 4469 | 122 | |
GenBank mRNA | AB208934.1 | - | 26 - 27 | 3162 | 122 |
AF030339.1 | - | 27 - 28 | 4481 | 122 | |
AK295226.1 | - | 12 - 13 | 1610 | 122 | |
AK307920.1 | - | 8 - 9 | 928 | 122 |