The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005116.5 | NP_005107.4 | 12 - 13 | 1622 | 140 |
NM_203327.1 | NP_976072.1 | 12 - 13 | 1640 | 140 | |
XM_011529414.2 | XP_011527716.1 | 12 - 13 | 1627 | 140 | |
XM_011529415.2 | XP_011527717.1 | 11 - 12 | 1448 | 140 | |
XM_011529416.2 | XP_011527718.1 | 11 - 12 | 1757 | 140 | |
XM_011529417.2 | XP_011527719.1 | 11 - 12 | 1504 | 140 | |
XM_017028171.1 | XP_016883660.1 | 12 - 13 | 1661 | 140 | |
XM_017028172.1 | XP_016883661.1 | 12 - 13 | 1665 | 140 | |
XM_017028173.1 | XP_016883662.1 | 12 - 13 | 1645 | 140 | |
XM_017028174.1 | XP_016883663.1 | 12 - 13 | 1540 | 140 | |
XM_017028175.1 | XP_016883664.1 | 8 - 9 | 976 | 140 | |
XM_017028176.1 | XP_016883665.1 | 9 - 10 | 1294 | 140 | |
XM_017028177.1 | XP_016883666.1 | 9 - 10 | 1281 | 140 | |
XM_017028178.1 | XP_016883667.1 | 11 - 12 | 1547 | 140 | |
XM_017028179.1 | XP_016883668.1 | 11 - 12 | 1534 | 140 | |
XM_017028180.1 | XP_016883669.1 | 7 - 8 | 858 | 140 | |
GenBank mRNA | AF058319.1 | - | 10 - 11 | 1247 | 140 |
AF092511.1 | - | 11 - 12 | 1543 | 140 | |
AF164142.1 | - | 12 - 13 | 1640 | 140 | |
AJ269478.1 | - | 10 - 11 | 1247 | 140 | |
AJ292318.1 | - | 11 - 12 | 1478 | 140 | |
AK296304.1 | - | 9 - 10 | 1288 | 140 | |
AY380556.1 | - | 10 - 11 | 1247 | 140 | |
D87075.2 | - | 12 - 13 | 1622 | 140 |