The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005116.5 | NP_005107.4 | 8 - 9 | 1019 | 98 |
NM_203327.1 | NP_976072.1 | 8 - 9 | 1037 | 98 | |
XM_011529414.2 | XP_011527716.1 | 8 - 9 | 1024 | 98 | |
XM_011529415.2 | XP_011527717.1 | 7 - 8 | 845 | 98 | |
XM_011529416.2 | XP_011527718.1 | 7 - 8 | 1154 | 98 | |
XM_011529417.2 | XP_011527719.1 | 7 - 8 | 901 | 98 | |
XM_017028171.1 | XP_016883660.1 | 8 - 9 | 1058 | 98 | |
XM_017028172.1 | XP_016883661.1 | 8 - 9 | 1062 | 98 | |
XM_017028173.1 | XP_016883662.1 | 8 - 9 | 1042 | 98 | |
XM_017028174.1 | XP_016883663.1 | 8 - 9 | 937 | 98 | |
XM_017028175.1 | XP_016883664.1 | 4 - 5 | 373 | 98 | |
XM_017028178.1 | XP_016883667.1 | 7 - 8 | 944 | 98 | |
XM_017028179.1 | XP_016883668.1 | 7 - 8 | 931 | 98 | |
XM_017028180.1 | XP_016883669.1 | 3 - 4 | 255 | 98 | |
XM_017028181.1 | XP_016883670.1 | 8 - 9 | 1036 | 98 | |
GenBank mRNA | AF058319.1 | - | 6 - 7 | 644 | 98 |
AF164142.1 | - | 8 - 9 | 1037 | 98 | |
AJ269478.1 | - | 6 - 7 | 644 | 98 | |
AJ292318.1 | - | 7 - 8 | 875 | 98 | |
AY380556.1 | - | 6 - 7 | 644 | 98 | |
BC013112.2 | - | 8 - 9 | 1008 | 98 | |
D87075.2 | - | 8 - 9 | 1019 | 98 | |
EF032501.1 | - | 6 - 7 | 644 | 98 |