The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024769.3 | NP_079045.1 | 2 - 3 | 544 | 94 |
XM_011542998.2 | XP_011541300.1 | 2 - 3 | 512 | 94 | |
GenBank mRNA | AK026068.1 | - | 2 - 3 | 520 | 94 |
AK304843.1 | - | 2 - 3 | 491 | 94 | |
AY326422.1 | - | 2 - 3 | 368 | 94 | |
AY358340.1 | - | 2 - 3 | 368 | 94 | |
BC009371.2 | - | 2 - 3 | 190 | 94 | |
BE272517.1 | - | 2 - 3 | 438 | 94 | |
CA395814.1 | - | 2 - 3 | 457 | 94 |