The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007187.3 | NP_009118.1 | 1 - 2 | 115 | 66 |
GenBank mRNA | AF071185.1 | - | 1 - 2 | 151 | 66 |
AK000979.1 | - | 1 - 2 | 167 | 66 | |
AK001557.1 | - | 1 - 2 | 150 | 66 | |
AK297536.1 | - | 1 - 2 | 116 | 66 | |
AK314525.1 | - | 1 - 2 | 152 | 66 | |
BC104879.1 | - | 1 - 2 | 59 | 66 | |
BC108310.1 | - | 1 - 2 | 116 | 66 |