The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006770.3 | NP_006761.1 | 2 - 3 | 337 | 118 |
XM_011512082.1 | XP_011510384.1 | 2 - 3 | 340 | 118 | |
XM_017005171.1 | XP_016860660.1 | 2 - 3 | 340 | 118 | |
GenBank mRNA | AF035819.1 | - | 2 - 3 | 205 | 118 |
AK297030.1 | - | 2 - 3 | 184 | 118 | |
AK301407.1 | - | 3 - 4 | 354 | 118 | |
AK313139.1 | - | 2 - 3 | 286 | 118 | |
AX429246.1 | - | 2 - 3 | 205 | 118 | |
BC016004.1 | - | 2 - 3 | 341 | 118 |