The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006770.3 | NP_006761.1 | 9 - 10 | 997 | 152 |
XM_011512082.1 | XP_011510384.1 | 9 - 10 | 1000 | 152 | |
XM_011512083.2 | XP_011510385.1 | 6 - 7 | 609 | 152 | |
GenBank mRNA | AF035819.1 | - | 9 - 10 | 865 | 152 |
AK297030.1 | - | 9 - 10 | 844 | 152 | |
AK301407.1 | - | 10 - 11 | 1014 | 152 | |
AK313139.1 | - | 9 - 10 | 946 | 152 | |
AX429246.1 | - | 9 - 10 | 865 | 152 | |
BC016004.1 | - | 9 - 10 | 1001 | 152 |