The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256213.1 | NP_001243142.1 | 11 - 12 | 1502 | 83 |
NM_001256214.1 | NP_001243143.1 | 11 - 12 | 1672 | 83 | |
NM_152296.4 | NP_689509.1 | 11 - 12 | 1633 | 83 | |
GenBank mRNA | AK094628.1 | - | 3 - 4 | 337 | 83 |
AK122693.1 | - | 5 - 6 | 755 | 83 | |
AK223569.1 | - | 11 - 12 | 1584 | 83 | |
AK293784.1 | - | 10 - 11 | 1284 | 83 | |
AK295078.1 | - | 11 - 12 | 1626 | 83 | |
AK295833.1 | - | 11 - 12 | 1487 | 83 | |
AK296557.1 | - | 11 - 12 | 1502 | 83 | |
AK307921.1 | - | 11 - 12 | 1580 | 83 | |
AK316069.1 | - | 11 - 12 | 1619 | 83 | |
BC009282.2 | - | 11 - 12 | 1549 | 83 | |
BC009394.2 | - | 11 - 12 | 1549 | 83 | |
BC015566.2 | - | 11 - 12 | 1549 | 83 |