The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033274.4 | NP_150377.1 | 8 - 9 | 872 | 69 |
XM_005266003.2 | XP_005266060.1 | 8 - 9 | 824 | 69 | |
GenBank mRNA | AF134707.1 | - | 5 - 6 | 453 | 69 |
AF311317.1 | - | 8 - 9 | 781 | 69 | |
AF326918.1 | - | 8 - 9 | 793 | 69 | |
AK308182.1 | - | 8 - 9 | 824 | 69 | |
Y13786.2 | - | 8 - 9 | 822 | 69 |