Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IKZF1
Entrez Gene ID
10320
Gene Name
IKAROS family zinc finger 1
Gene Aliases
CVID13, Hs.54452, IK1, IKAROS, LYF1, LyF-1, PPP1R92, PRO0758, ZNFN1A1
Chromosome Location
Chr.7: 50303453 - 50405101 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001220767.2 NP_001207696.1 4 - 5 467 91
NM_001291838.1 NP_001278767.1 5 - 6 694 91
NM_001291841.1 NP_001278770.1 3 - 4 299 91
NM_001291842.1 NP_001278771.1 3 - 4 299 91
NM_006060.5 NP_006051.1 6 - 7 982 91
XM_011515058.2 XP_011513360.1 7 - 8 872 91
XM_011515059.2 XP_011513361.1 7 - 8 874 91
XM_011515060.2 XP_011513362.1 7 - 8 4022 91
XM_011515061.2 XP_011513363.1 6 - 7 810 91
XM_011515062.2 XP_011513364.1 7 - 8 1152 91
XM_011515063.2 XP_011513365.1 7 - 8 1069 91
XM_011515064.2 XP_011513366.1 7 - 8 4700 91
XM_011515065.2 XP_011513367.1 7 - 8 837 91
XM_011515066.2 XP_011513368.1 7 - 8 941 91
XM_011515068.2 XP_011513370.1 6 - 7 1009 91
XM_011515069.2 XP_011513371.1 6 - 7 4637 91
XM_011515071.2 XP_011513373.1 6 - 7 611 91
XM_011515072.2 XP_011513374.1 5 - 6 549 91
XM_011515073.2 XP_011513375.1 6 - 7 891 91
XM_011515074.2 XP_011513376.1 5 - 6 552 91
XM_011515075.2 XP_011513377.1 5 - 6 831 91
XM_011515076.2 XP_011513378.1 5 - 6 775 91
XM_017011667.1 XP_016867156.1 6 - 7 808 91
XM_017011668.1 XP_016867157.1 5 - 6 515 91
XM_017011669.1 XP_016867158.1 6 - 7 777 91
XM_017011670.1 XP_016867159.1 6 - 7 1138 91
GenBank mRNA AK303586.1 - 5 - 6 653 91
U40462.1 - 6 - 7 882 91

View Full Product Details