Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ELAVL2
- Entrez Gene ID
-
1993
- Gene Name
- ELAV like neuron-specific RNA binding protein 2
- Gene Aliases
-
HEL-N1, HELN1, HUB
- Chromosome Location
-
Chr.9: 23690099 - 23850757 on Build GRCh38
View Full Product Details