The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057175.3 | NP_476516.1 | 7 - 8 | 1068 | 100 |
XM_005263236.2 | XP_005263293.1 | 7 - 8 | 1123 | 100 | |
GenBank mRNA | AF327722.1 | - | 7 - 8 | 1091 | 100 |
AJ314788.1 | - | 7 - 8 | 1020 | 100 | |
AK023402.1 | - | 7 - 8 | 1091 | 100 | |
AK304417.1 | - | 6 - 7 | 749 | 100 | |
AY039242.1 | - | 7 - 8 | 1093 | 100 | |
AY112670.1 | - | 7 - 8 | 812 | 100 | |
BC032642.1 | - | 7 - 8 | 1093 | 100 | |
BC039818.1 | - | 7 - 8 | 1083 | 100 | |
BC093928.1 | - | 7 - 8 | 877 | 100 | |
BC104806.1 | - | 7 - 8 | 877 | 100 |