The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016639.2 | NP_057723.1 | 2 - 3 | 285 | 102 |
GenBank mRNA | AB035480.1 | - | 2 - 3 | 238 | 102 |
AF191148.1 | - | 2 - 3 | 228 | 102 | |
BC002718.2 | - | 2 - 3 | 256 | 102 | |
BM842283.1 | - | 2 - 3 | 291 | 102 |