The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032287.2 | NP_115663.2 | 2 - 2 | 2606 | 98 |
GenBank mRNA | AK126304.1 | - | 1 - 1 | 2013 | 98 |
BC030232.1 | - | 2 - 2 | 2606 | 98 | |
BX648506.1 | - | 1 - 1 | 805 | 98 |