The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001220765.2 | NP_001207694.1 | 5 - 6 | 857 | 113 |
NM_001220770.2 | NP_001207699.1 | 3 - 4 | 342 | 113 | |
NM_001291837.1 | NP_001278766.1 | 5 - 6 | 830 | 113 | |
NM_001291839.1 | NP_001278768.1 | 4 - 5 | 374 | 113 | |
XM_011515067.2 | XP_011513369.1 | 6 - 7 | 749 | 113 | |
XM_011515070.2 | XP_011513372.1 | 5 - 6 | 688 | 113 | |
XM_011515077.2 | XP_011513379.1 | 4 - 5 | 427 | 113 | |
XM_011515078.2 | XP_011513380.1 | 4 - 5 | 430 | 113 | |
GenBank mRNA | AK308411.1 | - | 5 - 6 | 787 | 113 |
AY377974.1 | - | 5 - 6 | 757 | 113 | |
BC018349.1 | - | 5 - 6 | 781 | 113 | |
BT009836.1 | - | 5 - 6 | 589 | 113 | |
DC391353.1 | - | 4 - 5 | 380 | 113 |