Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAA15
Entrez Gene ID
80155
Gene Name
N(alpha)-acetyltransferase 15, NatA auxiliary subunit
Gene Aliases
Ga19, NARG1, NAT1P, NATH, TBDN, TBDN100
Chromosome Location
Chr.4: 139301467 - 139390781 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK023402.1 - 13 - 14 1815 145

View Full Product Details