Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2U1
Entrez Gene ID
113612
Gene Name
cytochrome P450 family 2 subfamily U member 1
Gene Aliases
P450TEC, SPG49, SPG56
Chromosome Location
Chr.4: 107931549 - 107953461 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183075.2 NP_898898.1 4 - 5 1541 91
XM_005262717.2 XP_005262774.1 4 - 5 1607 91
XM_005262720.2 XP_005262777.1 3 - 4 917 91
GenBank mRNA AK290002.1 - 4 - 5 1546 91
AK298989.1 - 3 - 4 463 91
AK303284.1 - 5 - 6 1346 91
AK304724.1 - 5 - 6 1419 91
AY343323.1 - 4 - 5 1497 91
BC132767.1 - 4 - 5 1506 91
BC136483.1 - 4 - 5 1506 91

View Full Product Details