The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007282.4 | NP_009213.1 | 8 - 9 | 1395 | 150 |
NM_183381.2 | NP_899237.1 | 7 - 8 | 1293 | 150 | |
XM_005247092.3 | XP_005247149.1 | 7 - 8 | 1252 | 150 | |
XM_011512373.2 | XP_011510675.1 | 9 - 10 | 1400 | 150 | |
XM_011512374.2 | XP_011510676.1 | 8 - 9 | 1298 | 150 | |
XM_011512376.2 | XP_011510678.1 | 7 - 8 | 703 | 150 | |
XM_017005654.1 | XP_016861143.1 | 8 - 9 | 1474 | 150 | |
XM_017005655.1 | XP_016861144.1 | 7 - 8 | 1375 | 150 | |
XM_017005656.1 | XP_016861145.1 | 8 - 9 | 1354 | 150 | |
XM_017005657.1 | XP_016861146.1 | 8 - 9 | 1487 | 150 | |
XM_017005658.1 | XP_016861147.1 | 7 - 8 | 1385 | 150 | |
XM_017005659.1 | XP_016861148.1 | 6 - 7 | 1122 | 150 | |
XM_017005660.1 | XP_016861149.1 | 6 - 7 | 1112 | 150 | |
GenBank mRNA | AF037204.1 | - | 7 - 8 | 762 | 150 |
AF070558.1 | - | 7 - 8 | 753 | 150 | |
AK090638.1 | - | 7 - 8 | 1273 | 150 | |
AK090771.1 | - | 4 - 5 | 382 | 150 | |
AK313304.1 | - | 7 - 8 | 777 | 150 | |
BC009781.1 | - | 7 - 8 | 770 | 150 | |
BC009803.1 | - | 8 - 9 | 873 | 150 | |
BC017878.1 | - | 7 - 8 | 735 | 150 | |
BM556246.1 | - | 6 - 7 | 648 | 150 | |
CB109037.1 | - | 1 - 2 | 120 | 150 | |
CR456804.1 | - | 6 - 7 | 611 | 150 |