Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRIQ3
Entrez Gene ID
127255
Gene Name
leucine rich repeats and IQ motif containing 3
Gene Aliases
LRRC44
Chromosome Location
Chr.1: 74026015 - 74198201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105659.1 NP_001099129.1 2 - 3 436 158
NM_001322315.1 NP_001309244.1 2 - 3 436 158
XM_011540663.2 XP_011538965.1 2 - 3 450 158
XM_017000286.1 XP_016855775.1 2 - 3 450 158
XM_017000287.1 XP_016855776.1 2 - 3 450 158
XM_017000288.1 XP_016855777.1 2 - 3 450 158
GenBank mRNA AK131476.2 - 2 - 3 393 158
BC020778.1 - 2 - 3 396 158

View Full Product Details