The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105659.1 | NP_001099129.1 | 6 - 7 | 1190 | 131 |
NM_001322315.1 | NP_001309244.1 | 6 - 7 | 1190 | 131 | |
XM_011540658.1 | XP_011538960.1 | 5 - 6 | 880 | 131 | |
XM_011540659.1 | XP_011538961.1 | 4 - 5 | 394 | 131 | |
XM_011540660.1 | XP_011538962.1 | 4 - 5 | 2519 | 131 | |
XM_011540662.2 | XP_011538964.1 | 3 - 4 | 818 | 131 | |
GenBank mRNA | AK131476.2 | - | 8 - 9 | 3512 | 131 |