Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRIQ3
Entrez Gene ID
127255
Gene Name
leucine rich repeats and IQ motif containing 3
Gene Aliases
LRRC44
Chromosome Location
Chr.1: 74026015 - 74198201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105659.1 NP_001099129.1 7 - 8 1906 160
XM_011540658.1 XP_011538960.1 6 - 7 1596 160
XM_011540659.1 XP_011538961.1 5 - 6 1110 160
XM_011540660.1 XP_011538962.1 5 - 6 3235 160
XM_011540662.2 XP_011538964.1 4 - 5 1534 160
GenBank mRNA BC062795.1 - 1 - 2 634 160

View Full Product Details