The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011552.3 | NP_001011552.2 | 1 - 2 | 715 | 89 |
XM_011511158.1 | XP_011509460.1 | 1 - 2 | 736 | 89 | |
GenBank mRNA | CR627411.1 | - | 1 - 2 | 716 | 89 |