Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ODF2
Entrez Gene ID
4957
Gene Name
outer dense fiber of sperm tails 2
Gene Aliases
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location
Chr.9: 128455155 - 128501292 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242352.1 NP_001229281.1 10 - 11 1364 120
NM_001242353.1 NP_001229282.1 10 - 11 1450 120
NM_001242354.1 NP_001229283.1 9 - 10 1136 120
NM_002540.4 NP_002531.3 12 - 13 1652 120
NM_153432.1 NP_702910.1 9 - 10 1210 120
NM_153433.1 NP_702911.1 10 - 11 1377 120
NM_153435.1 NP_702913.1 10 - 11 1270 120
NM_153436.1 NP_702914.1 9 - 10 1034 120
NM_153437.2 NP_702915.1 10 - 11 1300 120
NM_153439.1 NP_702917.1 9 - 10 1153 120
NM_153440.1 NP_702918.1 9 - 10 1080 120
XM_005252010.2 XP_005252067.1 9 - 10 1162 120
XM_005252011.2 XP_005252068.1 9 - 10 1102 120
XM_005252012.3 XP_005252069.1 10 - 11 1352 120
XM_005252013.3 XP_005252070.1 10 - 11 1299 120
XM_005252014.3 XP_005252071.1 10 - 11 1348 120
XM_005252017.4 XP_005252074.1 10 - 11 1253 120
XM_005252018.2 XP_005252075.1 11 - 12 1458 120
XM_005252019.3 XP_005252076.1 11 - 12 1477 120
XM_005252022.2 XP_005252079.1 10 - 11 1276 120
XM_005252023.2 XP_005252080.1 10 - 11 1247 120
XM_005252024.3 XP_005252081.1 10 - 11 1266 120
XM_005252026.4 XP_005252083.1 10 - 11 1195 120
XM_005252029.2 XP_005252086.1 10 - 11 1276 120
XM_006717126.3 XP_006717189.1 10 - 11 1352 120
XM_011518722.2 XP_011517024.1 10 - 11 1276 120
XM_011518723.2 XP_011517025.1 10 - 11 1219 120
XM_011518724.2 XP_011517026.1 11 - 12 1440 120
XM_011518725.2 XP_011517027.1 11 - 12 1444 120
XM_011518726.2 XP_011517028.1 11 - 12 1462 120
XM_011518728.2 XP_011517030.1 10 - 11 1213 120
XM_011518729.2 XP_011517031.1 10 - 11 1149 120
XM_011518730.2 XP_011517032.1 10 - 11 1276 120
XM_011518731.2 XP_011517033.1 9 - 10 1105 120
XM_011518732.2 XP_011517034.1 10 - 11 1062 120
XM_011518733.2 XP_011517035.1 9 - 10 1048 120
XM_011518744.1 XP_011517046.1 9 - 10 1102 120
XM_017014764.1 XP_016870253.1 11 - 12 1433 120
XM_017014765.1 XP_016870254.1 11 - 12 1452 120
XM_017014766.1 XP_016870255.1 9 - 10 1162 120
XM_017014767.1 XP_016870256.1 11 - 12 1444 120
XM_017014768.1 XP_016870257.1 10 - 11 1236 120
XM_017014769.1 XP_016870258.1 10 - 11 1265 120
XM_017014770.1 XP_016870259.1 11 - 12 1420 120
XM_017014771.1 XP_016870260.1 11 - 12 1401 120
XM_017014772.1 XP_016870261.1 11 - 12 1477 120
XM_017014773.1 XP_016870262.1 11 - 12 1453 120
XM_017014774.1 XP_016870263.1 10 - 11 1163 120
XM_017014775.1 XP_016870264.1 11 - 12 1420 120
XM_017014776.1 XP_016870265.1 11 - 12 1401 120
XM_017014777.1 XP_016870266.1 10 - 11 1203 120
XM_017014778.1 XP_016870267.1 10 - 11 1306 120
XM_017014779.1 XP_016870268.1 9 - 10 1047 120
XM_017014780.1 XP_016870269.1 10 - 11 1230 120
XM_017014781.1 XP_016870270.1 9 - 10 985 120
GenBank mRNA AF012549.1 - 10 - 11 1240 120
AF053970.1 - 8 - 9 939 120
AK093367.1 - 9 - 10 1160 120
AK299303.1 - 10 - 11 1212 120
AK300793.1 - 9 - 10 985 120
AK301754.1 - 9 - 10 1077 120
AK301842.1 - 9 - 10 1002 120
AK301855.1 - 9 - 10 1056 120
AK302684.1 - 10 - 11 1304 120
AK309917.1 - 10 - 11 1155 120
AY319414.1 - 9 - 10 1186 120
AY366499.1 - 10 - 11 1147 120
BC010629.1 - 12 - 13 1450 120
BC091500.1 - 10 - 11 1264 120
DQ444713.1 - 8 - 9 977 120
DQ444714.1 - 8 - 9 920 120

View Full Product Details