Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ODF2
- Entrez Gene ID
-
4957
- Gene Name
- outer dense fiber of sperm tails 2
- Gene Aliases
-
CT134, ODF2/1, ODF2/2, ODF84
- Chromosome Location
-
Chr.9: 128455155 - 128501292 on Build GRCh38
View Full Product Details