The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164317.1 | NP_001157789.1 | 45 - 46 | 7673 | 60 |
NM_001164318.1 | NP_001157790.1 | 44 - 45 | 7547 | 60 | |
NM_001164319.1 | NP_001157791.1 | 43 - 44 | 7508 | 60 | |
NM_001457.3 | NP_001448.2 | 44 - 45 | 7580 | 60 | |
XM_005264978.1 | XP_005265035.1 | 44 - 45 | 7581 | 60 | |
GenBank mRNA | AB191258.1 | - | 44 - 45 | 7525 | 60 |
AB209889.1 | - | 25 - 26 | 4449 | 60 | |
AB371580.1 | - | 45 - 46 | 7653 | 60 | |
AB371581.1 | - | 43 - 44 | 7486 | 60 | |
AB371582.1 | - | 44 - 45 | 7558 | 60 | |
AF042166.1 | - | 44 - 45 | 7546 | 60 | |
AF043045.1 | - | 44 - 45 | 7580 | 60 | |
AF238609.1 | - | 10 - 11 | 1625 | 60 | |
AL137574.1 | - | 11 - 12 | 1797 | 60 | |
BX641085.1 | - | 43 - 44 | 7105 | 60 | |
CR749793.1 | - | 42 - 43 | 6993 | 60 | |
M62994.1 | - | 1 - 2 | 203 | 60 |