The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277961.1 | NP_001264890.1 | 3 - 4 | 959 | 65 |
NM_001277962.1 | NP_001264891.1 | 3 - 4 | 959 | 65 | |
NM_003156.3 | NP_003147.2 | 3 - 4 | 959 | 65 | |
GenBank mRNA | AK314928.1 | - | 3 - 4 | 480 | 65 |
BC021300.2 | - | 3 - 4 | 951 | 65 | |
HQ317451.1 | - | 3 - 4 | 958 | 65 | |
JX014264.1 | - | 3 - 4 | 391 | 65 | |
U52426.1 | - | 3 - 4 | 956 | 65 |