Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STIM1
Entrez Gene ID
6786
Gene Name
stromal interaction molecule 1
Gene Aliases
D11S4896E, GOK, IMD10, STRMK, TAM, TAM1
Chromosome Location
Chr.11: 3855703 - 4093210 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277961.1 NP_001264890.1 4 - 5 1066 67
NM_001277962.1 NP_001264891.1 4 - 5 1066 67
NM_003156.3 NP_003147.2 4 - 5 1066 67
GenBank mRNA AK314928.1 - 4 - 5 587 67
BC021300.2 - 4 - 5 1058 67
HQ317451.1 - 4 - 5 1065 67
JX014264.1 - 4 - 5 498 67
U52426.1 - 4 - 5 1063 67

View Full Product Details