The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015150.1 | NP_055965.1 | 2 - 3 | 421 | 124 |
XM_005264985.1 | XP_005265042.1 | 2 - 3 | 378 | 124 | |
XM_005264986.2 | XP_005265043.1 | 2 - 3 | 275 | 124 | |
XM_006713069.1 | XP_006713132.1 | 2 - 3 | 284 | 124 | |
XM_011533530.1 | XP_011531832.1 | 2 - 3 | 446 | 124 | |
XM_017006007.1 | XP_016861496.1 | 2 - 3 | 405 | 124 | |
GenBank mRNA | AK074446.1 | - | 2 - 3 | 226 | 124 |
AU138240.1 | - | 2 - 3 | 482 | 125 | |
AY237654.1 | - | 2 - 3 | 412 | 124 | |
BC051336.1 | - | 2 - 3 | 198 | 124 | |
BC069209.1 | - | 2 - 3 | 198 | 124 | |
BC098270.1 | - | 2 - 3 | 245 | 124 | |
BC100667.1 | - | 2 - 3 | 245 | 124 | |
BC100669.1 | - | 2 - 3 | 245 | 124 | |
BC121795.1 | - | 2 - 3 | 245 | 124 | |
D42043.1 | - | 2 - 3 | 349 | 124 |