The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015150.1 | NP_055965.1 | 6 - 7 | 1315 | 108 |
XM_005264985.1 | XP_005265042.1 | 6 - 7 | 1272 | 108 | |
XM_005264986.2 | XP_005265043.1 | 6 - 7 | 1169 | 108 | |
XM_006713069.1 | XP_006713132.1 | 6 - 7 | 1178 | 108 | |
XM_011533529.1 | XP_011531831.1 | 8 - 9 | 1712 | 108 | |
XM_011533530.1 | XP_011531832.1 | 6 - 7 | 1340 | 108 | |
XM_017006005.1 | XP_016861494.1 | 8 - 9 | 1712 | 108 | |
GenBank mRNA | AK074446.1 | - | 6 - 7 | 1121 | 108 |
AK301024.1 | - | 5 - 6 | 1008 | 108 | |
AK303846.1 | - | 2 - 3 | 312 | 108 | |
AY237654.1 | - | 6 - 7 | 1306 | 108 | |
BC006400.2 | - | 2 - 3 | 243 | 108 | |
BC014032.2 | - | 2 - 3 | 243 | 108 | |
BC032349.1 | - | 5 - 6 | 988 | 108 | |
BC051336.1 | - | 6 - 7 | 1092 | 108 | |
BC069209.1 | - | 6 - 7 | 1092 | 108 | |
BC098270.1 | - | 6 - 7 | 1139 | 108 | |
BC100667.1 | - | 6 - 7 | 1139 | 108 | |
BC100669.1 | - | 6 - 7 | 1139 | 108 | |
BC121795.1 | - | 6 - 7 | 1139 | 108 | |
D42043.1 | - | 6 - 7 | 1243 | 108 |