Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RFTN1
Entrez Gene ID
23180
Gene Name
raftlin, lipid raft linker 1
Gene Aliases
MIG2, PIB10, PIG9, RAFTLIN
Chromosome Location
Chr.3: 16315845 - 16513715 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015150.1 NP_055965.1 7 - 8 1426 92
XM_005264985.1 XP_005265042.1 7 - 8 1383 92
XM_005264986.2 XP_005265043.1 7 - 8 1280 92
XM_006713069.1 XP_006713132.1 7 - 8 1289 92
XM_011533529.1 XP_011531831.1 9 - 10 1823 92
XM_011533530.1 XP_011531832.1 7 - 8 1451 92
XM_017006005.1 XP_016861494.1 9 - 10 1823 92
GenBank mRNA AK074446.1 - 7 - 8 1232 92
AK301024.1 - 6 - 7 1119 92
AK303846.1 - 3 - 4 423 92
AY237654.1 - 7 - 8 1417 92
BC006400.2 - 3 - 4 354 92
BC014032.2 - 3 - 4 354 92
BC032349.1 - 6 - 7 1099 92
BC051336.1 - 7 - 8 1203 92
BC069209.1 - 7 - 8 1203 92
BC098270.1 - 7 - 8 1250 92
BC100667.1 - 7 - 8 1250 92
BC100669.1 - 7 - 8 1250 92
BC121795.1 - 7 - 8 1250 92
D42043.1 - 7 - 8 1354 92

View Full Product Details