The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134398.1 | NP_001127870.1 | 4 - 5 | 496 | 92 |
NM_003371.3 | NP_003362.2 | 4 - 5 | 496 | 92 | |
XM_005272213.1 | XP_005272270.1 | 4 - 5 | 546 | 92 | |
XM_017015108.1 | XP_016870597.1 | 4 - 5 | 546 | 92 | |
XM_017015109.1 | XP_016870598.1 | 4 - 5 | 546 | 92 | |
XM_017015110.1 | XP_016870599.1 | 4 - 5 | 546 | 92 | |
XM_017015111.1 | XP_016870600.1 | 4 - 5 | 546 | 92 | |
XM_017015112.1 | XP_016870601.1 | 4 - 5 | 546 | 92 | |
XM_017015113.1 | XP_016870602.1 | 4 - 5 | 546 | 92 | |
GenBank mRNA | BC132965.1 | - | 4 - 5 | 496 | 92 |
BC132967.1 | - | 4 - 5 | 496 | 92 | |
BX640754.1 | - | 4 - 5 | 336 | 92 | |
S76992.1 | - | 4 - 5 | 455 | 92 |