Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FXYD1
Entrez Gene ID
5348
Gene Name
FXYD domain containing ion transport regulator 1
Gene Aliases
PLM
Chromosome Location
Chr.19: 35137207 - 35143055 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278717.1 NP_001265646.1 2 - 3 308 64
NM_001278718.1 NP_001265647.1 2 - 3 202 64
NM_005031.4 NP_005022.2 2 - 3 198 64
NM_021902.3 NP_068702.1 2 - 3 174 64
XM_017026874.1 XP_016882363.1 2 - 3 153 64
XM_017026875.1 XP_016882364.1 2 - 3 368 64
XM_017026876.1 XP_016882365.1 2 - 3 303 64
GenBank mRNA AK289811.1 - 2 - 3 141 64
BC032800.2 - 1 - 2 68 64
BM691130.1 - 2 - 3 141 64
BM856555.1 - 2 - 3 135 64
BX115036.1 - 2 - 3 198 64
GQ258667.1 - 1 - 2 46 64
H57207.1 - 2 - 3 198 64
HY107209.1 - 2 - 3 175 64
HY111815.1 - 2 - 3 202 64
HY151101.1 - 2 - 3 163 64
HY151827.1 - 2 - 3 310 64
R46545.1 - 2 - 3 215 64
R72515.1 - 2 - 3 138 64
U72245.1 - 1 - 2 64 64

View Full Product Details