The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005415.4 | NP_005406.3 | 7 - 8 | 1589 | 72 |
XM_017004768.1 | XP_016860257.1 | 7 - 8 | 1338 | 72 | |
XM_017004769.1 | XP_016860258.1 | 8 - 9 | 3816 | 72 | |
XM_017004770.1 | XP_016860259.1 | 8 - 9 | 1619 | 72 | |
XM_017004771.1 | XP_016860260.1 | 9 - 10 | 1655 | 72 | |
GenBank mRNA | AK123420.1 | - | 2 - 3 | 1127 | 72 |
AK312738.1 | - | 7 - 8 | 1508 | 72 | |
BC019944.1 | - | 7 - 8 | 1496 | 72 | |
BC075818.1 | - | 1 - 2 | 98 | 72 | |
L20859.1 | - | 7 - 8 | 1420 | 72 |