Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLIN5
Entrez Gene ID
440503
Gene Name
perilipin 5
Gene Aliases
LSDA5, LSDP5, MLDP, OXPAT
Chromosome Location
Chr.19: 4522531 - 4535196 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013706.2 NP_001013728.2 4 - 5 392 81
GenBank mRNA BC033570.1 - 4 - 5 392 81
BC131524.1 - 4 - 5 389 81
DQ839131.1 - 4 - 5 387 81

View Full Product Details