Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NNT
Entrez Gene ID
23530
Gene Name
nicotinamide nucleotide transhydrogenase
Gene Aliases
GCCD4
Chromosome Location
Chr.5: 43602574 - 43705566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012343.3 NP_036475.3 15 - 16 2547 85
NM_182977.2 NP_892022.2 15 - 16 2513 85
XM_005248274.4 XP_005248331.1 15 - 16 2525 85
XM_005248275.4 XP_005248332.1 10 - 11 1775 85
XM_006714461.3 XP_006714524.1 14 - 15 2139 85
XM_011514001.2 XP_011512303.1 15 - 16 2544 85
XM_011514002.2 XP_011512304.1 14 - 15 2234 85
XM_017009293.1 XP_016864782.1 15 - 16 2415 85
GenBank mRNA AK297558.1 - 8 - 9 1160 85
AK299622.1 - 7 - 8 977 85
AL831822.2 - 15 - 16 2514 85
BC110543.1 - 15 - 16 2363 85
BC110544.1 - 14 - 15 2360 85
U40490.1 - 15 - 16 2435 85
Z50101.1 - 14 - 15 2335 85

View Full Product Details