The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012343.3 | NP_036475.3 | 1 - 2 | 204 | 105 |
GenBank mRNA | AK297558.1 | - | 1 - 2 | 204 | 105 |
AL831822.2 | - | 1 - 2 | 171 | 105 | |
DA521839.1 | - | 1 - 2 | 204 | 105 |