Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NNT
Entrez Gene ID
23530
Gene Name
nicotinamide nucleotide transhydrogenase
Gene Aliases
GCCD4
Chromosome Location
Chr.5: 43602574 - 43705566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012343.3 NP_036475.3 4 - 5 853 104
NM_182977.2 NP_892022.2 4 - 5 819 104
XM_005248274.4 XP_005248331.1 4 - 5 831 104
XM_006714461.3 XP_006714524.1 3 - 4 445 104
XM_011514001.2 XP_011512303.1 4 - 5 850 104
XM_011514002.2 XP_011512304.1 3 - 4 540 104
XM_017009293.1 XP_016864782.1 4 - 5 721 104
GenBank mRNA AL831822.2 - 4 - 5 820 104
BC110543.1 - 4 - 5 669 104
BC110544.1 - 3 - 4 666 104
U40490.1 - 4 - 5 741 104
Z50101.1 - 3 - 4 641 104

View Full Product Details