The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308101.1 | NP_001295030.1 | 5 - 6 | 738 | 148 |
NM_020640.3 | NP_065691.2 | 5 - 6 | 720 | 148 | |
XM_011512912.1 | XP_011511214.1 | 5 - 6 | 689 | 148 | |
XM_011512914.1 | XP_011511216.1 | 5 - 6 | 728 | 148 | |
XM_011512915.2 | XP_011511217.1 | 6 - 7 | 1250 | 148 | |
XM_011512916.2 | XP_011511218.1 | 5 - 6 | 1178 | 148 | |
XM_017006633.1 | XP_016862122.1 | 4 - 5 | 605 | 148 | |
XM_017006634.1 | XP_016862123.1 | 5 - 6 | 694 | 148 | |
XM_017006635.1 | XP_016862124.1 | 6 - 7 | 1250 | 148 | |
GenBank mRNA | AF292100.2 | - | 5 - 6 | 637 | 148 |
AF456425.1 | - | 5 - 6 | 623 | 148 | |
AF456426.1 | - | 5 - 6 | 623 | 148 | |
AK025764.1 | - | 3 - 4 | 283 | 148 | |
AK056335.1 | - | 5 - 6 | 653 | 148 | |
AK297328.1 | - | 6 - 7 | 537 | 148 | |
AK314480.1 | - | 5 - 6 | 637 | 148 | |
BC009478.2 | - | 5 - 6 | 642 | 148 | |
BC013163.1 | - | 4 - 5 | 593 | 148 |