The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015268.3 | NP_056083.3 | 20 - 21 | 2263 | 70 |
XM_017006036.1 | XP_016861525.1 | 7 - 8 | 720 | 70 | |
GenBank mRNA | AB014578.2 | - | 20 - 21 | 2312 | 70 |
AK127112.1 | - | 4 - 5 | 351 | 70 | |
AY369172.1 | - | 2 - 3 | 185 | 70 | |
AY779857.1 | - | 20 - 21 | 2263 | 70 | |
BC151246.1 | - | 21 - 22 | 2333 | 70 |