The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015268.3 | NP_056083.3 | 31 - 32 | 3535 | 88 |
XM_005247245.4 | XP_005247302.1 | 32 - 33 | 3732 | 88 | |
XM_017006036.1 | XP_016861525.1 | 18 - 19 | 1992 | 88 | |
GenBank mRNA | AB014578.2 | - | 31 - 32 | 3584 | 88 |
AK125330.1 | - | 8 - 9 | 919 | 88 | |
AY779857.1 | - | 31 - 32 | 3535 | 88 | |
BC151246.1 | - | 32 - 33 | 3605 | 88 |